Stampa…

Anti-BDNF (35928.11) Mouse Monoclonal Antibody

Fornitore: Merck Millipore (Oncogene)
GF35L-100
GF35L-100EA 680 EUR
GF35L-100
Anti-BDNF (35928.11) Mouse Monoclonal Antibody
Anticorpi
Type: Primary
Antigen: BDNF (35928.11)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR