Stampa…
Flow Cells for MinION™ and GridION™ Devices
Fornitore: Oxford Nanopore Technologies
FLO-MIN114 FLO-MIN114.8 FLOMIN004RAA.8 FLOMIN04RAA848 FLOMIN114.8.48 FLOMIN114.8.96 FLOMIN04RAA896 FLO-MIN004RAA
FLO-MIN114EA 700 EUR
FLO-MIN114 FLO-MIN114.8 OXNTFLOMIN004RAA.8 OXNTFLOMIN04RAA848 OXNTFLOMIN114.8.48 OXNTFLOMIN114.8.96 OXNTFLOMIN04RAA896 FLO-MIN004RAA
Flow Cells for MinION™ and GridION™ Devices
Celle a flusso
Nanopore sequencing flow cells for use with MinION™ and GridION™ devices.
Flow cells contain the proprietary sensor array, Application-Specific Integrated Circuit (ASIC), and nanopores for DNA/RNA sequencing on MinION™ and GridION™ devices.
R10.4.1 flow cells contain a double reader-head and are suitable for experiments where high consensus accuracy is required. Compatible with Kit 14 chemistry, R10.4.1 flow cells generate data at a modal accuracy above 99%. R9.4.1 flow cells are compatible with the previous generation of nanopore sequencing chemistries. These flow cells and associatedMore Product Information
Visualizza i codici
Please note that Oxford Nanopore Technologies requires the acceptance of their Terms & Conditions prior to product order placement. If you select product(s) to order, the Terms and Conditions will be displayed in the shopping cart for your review and acceptance.