Stampa…

Fucsina (acido) for tissue, stain, Sigma-Aldrich®

Fornitore: SIGMA ALDRICH MICROSCOPY


M-Clarity™ quality level = MQ200

F8129-100G F8129-25G F8129-50G F8129-100G F8129-25G F8129-50G
F8129-100GEA 337 EUR
F8129-100G F8129-25G F8129-50G SIALF8129-100G SIALF8129-25G SIALF8129-50G
Fucsina (acido) for tissue, stain, Sigma-Aldrich®
Fucsina (acido)

Acid Fuchsin is mainly used as a diachrome and is part of the connective tissue stains. It also works as a pH indicator.


  • Acid Fuchsin has been used in tissue sections for Masson's trichrome staining
  • It has been used as a marker to trace ingestion of solutions by green peach aphids

Formula: C₂₀H₁₇N₃Na₂O₉S₃
Punto di fusione: > 130 °C
Temperatura di stoccaggio: Room Temperature
MDL Number: MFCD00013286
CAS Number: 3244-88-0
EINECS: 221-816-5
Merck Index: 12,00109
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4018bf04429ebc2757c43d7c7a9de213ceef8186&vid=3dfb10465fa2d34a2f4b414e7f6e92b1f7ea2f15&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance (Form) Powder
Solubility (Colour) Red
Solubility (Turbidity)
4 mg + 4 ml, H₂O
Clear
Nitrogen 4.3 - 7.2 %
Recommended Retest Period 4 years
% Dye content
(approx. 70%)
≥60 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR