Stampa…
pH combination electrode, Eutech
Fornitore: EUTECH
ECFE7352801B
EUTEECFE7352801BEA 203 EUR
EUTEECFE7352801B
pH combination electrode, Eutech
Elettrodi Elettrodi pH
General purpose "3-in-1" pH/temperature combination electrode for use with CyberScan models (pH 10 / pH 100 / pH 200 / pH 500 / pH 1000 / pH 2000).