Stampa…
Folding magnifiers, Mobilent LED
Fornitore: ESCHENBACH OPTIK
1520910 152094 152097
ESCH1520910EA 58.7 EUR
ESCH1520910 ESCH152094 ESCH152097
Folding magnifiers, Mobilent LED
Lenti d'ingrandimento
Very compact, high magnification folding magnifier with efficient LED illumination.
  • Small size and handy for wearing around the neck
  • Battery cover thread in when using the lanyard
  • Plastic case housing thightly holding the lens
Suitable for on the go.
Informazioni per l'ordine: Supplied with a lanyard and CR 2032 battery.