Stampa…
Cleanroom anti-fatigue mats, Ergomat® AFS Complete
Fornitore: ERGOMAT
STF0203 STF0304 SX0412 SX0406 SX0320 SX0306 SX0304 SX0220 SX0209 SX0206 SX0203 STF0420 STF0412 STF0406 STF0306 STF0220 STF0209 STF0206 STF0320 SX0420 SX0204 SX0212 STF0312 STF0404 STF0409 SX0409 SX0303 SX0309 SX0312 SX0404 STF0309 STF0204 STF0303 STF0212
115-0958EA 149 EUR
115-0958 115-0962 115-0976 115-0975 115-0974 115-0973 115-0972 115-0971 115-0970 115-0969 115-0968 115-0967 115-0966 115-0965 115-0963 115-0961 115-0960 115-0959 115-0964 115-0977 ERGOSX0204 ERGOSX0212 ERGOSTF0312 ERGOSTF0404 ERGOSTF0409 ERGOSX0409 ERGOSX0303 ERGOSX0309 ERGOSX0312 ERGOSX0404 ERGOSTF0309 ERGOSTF0204 ERGOSTF0303 ERGOSTF0212
Cleanroom anti-fatigue mats, Ergomat® AFS Complete
Tappetini Tappetini antifatica
The anti-static Ergomat® AFS Complete is made of pure polyurethane and is designed to increase comfort while reducing trip hazards and body pain. This mat is for all dry and damp areas where flame retardant and anti-static material is important. Great chemical resistancy for where chemical or oil spillage may occur. The smooth version is ideal for areas where trolleys and carts are used.
  • Choice of smooth or bubbled upper surface
  • Fire retardant, resists moderate oils and liquids
  • Moulded bevelled edges prevent curling, eliminates trip hazards
  • Raw virgin