Stampa…
Human recombinant yellow fever NS1
Fornitore: ENZO LIFE SCIENCES
ENZPRT1090100
ENZOENZPRT1090100EA 482 EUR
ENZOENZPRT1090100
Human recombinant yellow fever NS1
Proteine e Peptidi
Yellow Fever virus NS1 protein presented in its native folded state with all post-translational modifications to deliver optimal antigenicity.
Produced in HEK 293 cells. Yellow Fever Virus NS1 protein from strain 17D.