Stampa…
PCR decontamination kit
Fornitore: ENZO LIFE SCIENCES
ENZKIT1370100
ENZOENZKIT1370100EA 183 EUR
ENZOENZKIT1370100
PCR decontamination kit
Reagenti per acidi nucleici Modificazione ed etichettatura degli acidi nucleici
Rapid removal of contaminating DNA in PCR mastermixes, without reduction in PCR sensitivity.
  • Removes DNA contamination from PCR mastermixes and primers prior to PCR
  • Improves target detection by reducing background
  • Does not effect PCR sensitivity
  • Optimized for PCR and probe based qPCR mixes
The PCR Decontamination Kit uses a dsDNase to remove contaminating DNA from mastermixes. The double-strand specific property allows decontamination in the presence of primers and probe.The dsDNase is irreversibly inactivated by heating to 60 °C in the presence of DTT, ensuring that DNA template added to the mastermix after inactivation will not be digested. The mastermix can be used immediately after decontamination to run PCR reactions.
AMPIGENE® Taq polymerases have been treated to remove host DNA and additional clean-up is not necessary.
Informazioni per l'ordine: Contains Heat-labile dsDNase, DTT.