Stampa…
Nona-Ubiquitin (linear) (from E. coli)
Fornitore: ENZO LIFE SCIENCES
BMLUW08100100
ENZOBMLUW08100100EA 138 EUR
ENZOBMLUW08100100
Nona-Ubiquitin (linear) (from E. coli)
Proteine e Peptidi
Produced in E. coli.
Linear polyubiquitin chains are useful tools for investigating, amongst other things, the specificity and reactivity of deubiquitinylating enzymes (DUBs) and the recognition and interaction of linear polyubiquitin modified proteins with by ubiquitin binding domain (UBDs) containing proteins.