Stampa…
Hepta-Ubiquitin (linear) (from E. coli)
Fornitore: ENZO LIFE SCIENCES
BMLUW08000100
ENZOBMLUW08000100EA 138 EUR
ENZOBMLUW08000100
Hepta-Ubiquitin (linear) (from E. coli)
Proteine e Peptidi
Linear polyubiquitin chains are useful tools for investigating, amongst other things, the specificity and reactivity of deubiquitinylating enzymes (DUBs) and the recognition and interaction of linear polyubiquitin modified proteins with by ubiquitin binding domain (UBDs) containing proteins.