Stampa…
Human recombinant MMP-11 (catalytic domain) (from E. coli)
Fornitore: ENZO LIFE SCIENCES
BMLSE2820010
ENZOBMLSE2820010EA 475 EUR
ENZOBMLSE2820010
Human recombinant MMP-11 (catalytic domain) (from E. coli)
Proteine e Peptidi
Produced in E. coli. Active Matrix Metalloproteinase-11 (MMP-11, Stromelysin-3) catalytic domain from human cDNA.
  • Consists of the catalytic domain of human MMP-11 (Phe98-Ser266, NM_005940) with a C-terminal purification tag
MMPs lacking this domain cannot cleave native collagens; however, activity toward other targets such as gelatin, casein, or peptide substrates is unaffected. It may be an important link between obesity and cancer.
New Product
Visualizza i codici
Specifiche
Protein/peptide name MMP-11 (catalytic domain)
Environmentally Preferable
Protein/peptide type Recombinant
Specie Human
Sorgente luminosa E. coli