Stampa…
CinnGEL 2-methylester
Fornitore: ENZO LIFE SCIENCES
BMLPR1150001
ENZOBMLPR1150001EA 164 EUR
ENZOBMLPR1150001
CinnGEL 2-methylester
Proteine e Peptidi
PTP1B inhibitor
Novel peptoid inhibitor of PTP1B (IC50=1,3 µM). Methyl ester (for cell permeability) is hydrolyzed to active inhibitor by intracellular esterases.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=63ea2ad0051d56a4c93e3c5d7ce6a7ae8804725d&vid=46c3db5a561cf2395d6f5447af52a09e2f3fe069&prid=web&clid=SAPDAM
New Product
Visualizza i codici
Specifiche
Protein/peptide name CinnGEL 2-methylester
Environmentally Preferable
Specie
Sorgente luminosa