Stampa…
RR-src peptide
Fornitore: ENZO LIFE SCIENCES
BMLP3080001
ENZOBMLP3080001EA 136 EUR
ENZOBMLP3080001
RR-src peptide
Proteine e Peptidi
Src kinase substrate
This tyrosine kinase substrate sequence, RRLIEDAEYAARG, was derived from the v-Src autophosphorylation site (original src sequence is IEDNEYTARQG). Km=5 mM in LSTRA cell extracts.