Stampa…

Leukotriene C4 ≥95% (by HPLC)

Fornitore: ENZO LIFE SCIENCES
Pericolo

BMLLC0040050
ENZOBMLLC0040050EA 424 EUR
ENZOBMLLC0040050
Leukotriene C4 ≥95% (by HPLC)
Leukotriene C4

Prostanoid receptor ligand.


A cysteinyl leukotriene which acts as potent agonist for contraction of smooth muscle. Induces microvascular permeability and mucus secretion in airways. Enhances growth of human airway epithelial cells. Reduces myocardial contractility and coronary blood flow. Induces long lasting hypotension. Stimulates angiogenesis. Stimulates LH8, LHRH9 and prolactin release. Produced in specific cell types or by transcellular biosynthesis and rapidly metabolized to LTD4 and LTE4. Transported by the multidrug resistance protein (MRP; Km=35 nM).

Formula: C₃₀H₄₇N₃O₉S
Peso molecolare: 625.8 g/mol
Temperatura di stoccaggio: Ultracongelamento (–80°C)
CAS Number: 72025-60-6
UN: 1230
ADR: 3 II
Merck Index: 14,05455
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ac510bf624e831654bbc9a9d8bbc9e77ada9908d&vid=b0e9232b20afef6ce389d5704fd970cb43b5d43d&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Purity ≥95% (HPLC)
Concentration 50 µg/ml
Source Synthetic


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR