Stampa…

Anti-PTPRC Mouse Monoclonal Antibody

Fornitore: ENZO LIFE SCIENCES
ALX805041C100 ALX805041C100
ALEXALX805041C100EA 0 EUR
ALEXALX805041C100 ENZOALX805041C100
Anti-PTPRC Mouse Monoclonal Antibody
Anticorpi
Host: Mouse, Isotype: IgG1

  • For Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot

Recognizes all isoforms of human CD45.

Type: Primary
Antigen: PTPRC
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR