Stampa…

Anti-CCL2 Mouse Monoclonal Antibody

Fornitore: ENZO LIFE SCIENCES
ALX804465C100 ALX804465C100
ALEXALX804465C100EA 0 EUR
ALEXALX804465C100 ENZOALX804465C100
Anti-CCL2 Mouse Monoclonal Antibody
Anticorpi
Host: Mouse, Isotype: IgG1

  • For ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot

Recognizes human CCL2.

Type: Primary
Antigen: CCL2
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR