Stampa…

Anti-MAPK Mouse Monoclonal Antibody [clone: 12D4]

Fornitore: ENZO LIFE SCIENCES
ALX804061C100 ALX804061C100
ALEXALX804061C100EA 0 EUR
ALEXALX804061C100 ENZOALX804061C100
Anti-MAPK Mouse Monoclonal Antibody [clone: 12D4]
Anticorpi
Host: Mouse, Isotype: IgG1

  • For ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot

Recognizes the pThr-Glu-pTyr motif of activated human, mouse, rat and dog ERK1 and ERK2. Requires phosphorylation both at the threonine and the tyrosine site and does not cross-react with the non-phosphorylated form of the protein. Does not cross-react with activated JNK or p38.

Reconstitute with 1ml water (15 minutes at room temperature).

Type: Primary
Antigen: MAPK
Clonality: Monoclonal
Clone: 12D4
Conjugation:
Epitope:More Product Information
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR