Stampa…
Mouse recombinant TL1A (soluble)
Fornitore: ENZO LIFE SCIENCES
ALX522105C010
ENZOALX522105C010EA 618 EUR
ENZOALX522105C010
Mouse recombinant TL1A (soluble)
Proteine e Peptidi
Produced in HEK 293 cells. The extracellular domain of mouse TL1A (aa 94-252) is fused at the N-terminus to a linker peptide (8 aa) and a FLAG®-tag.