Stampa…

Rifampicin

Fornitore: ENZO LIFE SCIENCES

ALX380071M100
ENZOALX380071M100EA 51.7 EUR
ENZOALX380071M100
Rifampicin
Rifampicin
Formula: C₄₃H₅₈N₄O₁₂
Punto di fusione: 183…188 °C
MDL Number: MFCD00151389
CAS Number: 13292-46-1
EINECS: 236-312-0
Merck Index: 12,08382

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR