Stampa…

Cholesterol-3-sulphate sodium salt

Fornitore: ENZO LIFE SCIENCES

Synonyms: Cholesteryl sodium sulphate, Cholesteryl sulphate sodium

ALX370009M005
ENZOALX370009M005EA 50.6 EUR
ENZOALX370009M005
Cholesterol-3-sulphate sodium salt
Cholesterol-3-sulphate sodium salt

PKC activator.


Activates multiple protein kinase C (PKC) iso-enzymes, especially the ε, η and ζ isoforms.


Soluble in methanol.

Formula: C₂₇H₄₅O₄S·Na
Peso molecolare: 488.7 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00079091
CAS Number: 2864-50-8

Visualizza i codici

Risultati del test di specificazione

Purity One spot by TLC
Identity Determined by IR


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR