Stampa…
Bovine native PARG (from Thymus)
Fornitore: ENZO LIFE SCIENCES
ALX202045UC01 ALX202045UC01
ALEXALX202045UC01EA 0 EUR
ALEXALX202045UC01 ENZOALX202045UC01
Bovine native PARG (from Thymus)
Proteine e Peptidi
Isolated from calf thymus.
New Product
Visualizza i codici
Specifiche
Protein/peptide name PARG
Environmentally Preferable
Protein/peptide type Native
Specie Bovine
Sorgente luminosa Thymus
Purezza Purified by substrate chromatography. The enzyme is composed of two major bands at 74kDa and 59kDa
Formulazione Liquid. In 50 mM K-PO4 buffer, pH 7,5, containing 50 mM potassium chloride, 0,1% Triton-x 100, 10% glycerol and 1 mM DTT.
Note: add 10 mM beta-Mercaptoethanol.