Stampa…
Human recombinant hepsin
Fornitore: ENZO LIFE SCIENCES
ALX2017501
ENZOALX2017501EA 519 EUR
ENZOALX2017501
Human recombinant hepsin
Proteine e Peptidi
Produced in Drosophila Schneider 2 (S2) cells. Recombinant human hepsin (aa 45-417) is fused at the C-terminus to a V5-His-tag.