Stampa…
NBD peptide
Fornitore: ENZO LIFE SCIENCES
ALX1630111 ALX1630111
ALEXALX1630111EA 0 EUR
ALEXALX1630111 ENZOALX1630111
NBD peptide
Proteine e Peptidi
For increased cell permeability the peptide was covalently linked to the 10 aa recognized by the TAT transporter.
Informazioni per l'ordine: Quantity: 50 µl