Stampa…
Ac-muramyl-Ala-D-Glu-amide
Fornitore: ENZO LIFE SCIENCES
ALX151035M005
ENZOALX151035M005EA 253 EUR
ENZOALX151035M005
Ac-muramyl-Ala-D-Glu-amide
Proteine e Peptidi
NALP3 ligand
Specific ligand for NALP3. For inactive control compound see MDP-DD (Prod. No. ALX-151-036).