Stampa…
[pSer15]HSP27 (human), ELISA kit
Fornitore: ENZO LIFE SCIENCES
ADI900170
ENZOADI900170EA 764 EUR
ENZOADI900170
[pSer15]HSP27 (human), ELISA kit
Saggi ELISA
Only commercially available ELISA kit for ultra-sensitive detection of human phospho-Ser15 HSP27 in cell lysates, plasma, and serum.
For the quantitative determination of human [pSer15]HSP27 in cell lysates, plasma, and serum.
The [pSer15]HSP27 (human), EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in 3 hours. Save time, money, and precious sample with fully quantitative results and increased sensitivity compared to Western blot analysis.
Informazioni per l'ordine: Contains Microtiter plate, Conjugate, Antibody, Assay buffer 27, Wash buffer concentrate, Standard, TMB Substrate, 5X Extraction reagent, Stop solution 2.