Stampa…
11-dehydro-TXB2 ELISA Kits
Fornitore: ENZO LIFE SCIENCES
ADI900092
ENZOADI900092EA 527 EUR
ENZOADI900092
11-dehydro-TXB2 ELISA Kits
Saggi ELISA
For the quantitative determination of 11-dehydro-TXB2 in culture supernatants and urine from any species. Cited samples include serum.
The 11-dehydro-TXB2 EIA kit is a colorimetric competitive enzyme immunoassay kit with results in 3 hours. Absorbance is read at 405 nm. This kit offers ultrasensitive quantification of 11-d-TXB2 in a variety of sample matrices with negligible reactivity with related molecules in a convenient 96-well format offers higher throughput analysis compared to conventional analytical methods.
Informazioni per l'ordine: Contains GxR IgG Microtiter plate, Assay buffer, Conjugate, Antibody, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution. Alternative size available: 89141-324 (5x96 wells)