Stampa…
Pharmsteri™ PES Cartridge Filter
Fornitore: ENTEGRIS
PSTS045510S1 PSTS045710S1
ENTEPSTS045510S1EA 0 EUR
ENTEPSTS045510S1 ENTEPSTS045710S1
Pharmsteri™ PES Cartridge Filter
Filtri Cartucce con filtro
Pharmsteri PES cartridge filters help ensure reliable product processing in pharmaceutical applications.
  • Meets the requirements of WFI set by USP
  • Meets the criteria for a non-fiber releasing filter defined in 21 CFR 210.3(b)(6)
  • Tested according to USP<665>
  • Meets the FDA Indirect Food Additive requirements cited in 21 CFR 177-182
  • 20 cycles of steam-in-place (SIP) sterilization or autoclave, 30 min at 127 °C (261 °F)
Polypropylene components and polyethersulphone (PES) membrane construction allow use in a broad range of applications. OptimizedMore Product Information