Stampa…
Integra® Pneumatic Valves, 12,7 mm Orifice, 3-way
Fornitore: ENTEGRIS
D8-3C-8S3 D8-3C-8F-3 D8-3C-12F D8-3C-12F-3 D8-3C-12N D8-3C-12P D8-3C-12S1 D8-3C-12S3 D8-3C-8F D8-3C-8N D8-3C-8S1 D8-3C-8S2 D8-3C-8F-OM
ENTED8-3C-8S3EA 0 EUR
ENTED8-3C-8S3 ENTED8-3C-8F-3 ENTED8-3C-12F ENTED8-3C-12F-3 ENTED8-3C-12N ENTED8-3C-12P ENTED8-3C-12S1 ENTED8-3C-12S3 ENTED8-3C-8F ENTED8-3C-8N ENTED8-3C-8S1 ENTED8-3C-8S2 ENTED8-3C-8F-OM
Integra® Pneumatic Valves, 12,7 mm Orifice, 3-way
Valvole Valvole ellittiche
This high purity diversion valve minimises footprint chemical applications including CMP slurry.
  • All wetted parts are PFA providing chemical resistance in high purity, corrosive chemical applications
  • No metals or elastomers in secondary containment area improves reliability and cleanliness
PVDF actuator components and no external metal parts means lower contamination potential in the event of a media leak or exposure to harsh external environments.
Ideal in point-of-use applications when low flow is required, including high-purity, corrosiveMore Product Information