Stampa…
Cartridge filters, Etchgard™ HP and HPX
Fornitore: ENTEGRIS
CPUV01P01 CPUV01P01K CPUV01PLT CPUV01PLTK CPUV0SP01 CPUZ01P01 CPUZ01P01K CPUZ01PLT CPUZ01PLTK CPUZ0SP01
ENTECPUV01P01EA 0 EUR
ENTECPUV01P01 ENTECPUV01P01K ENTECPUV01PLT ENTECPUV01PLTK ENTECPUV0SP01 ENTECPUZ01P01 ENTECPUZ01P01K ENTECPUZ01PLT ENTECPUZ01PLTK ENTECPUZ0SP01
Cartridge filters, Etchgard™ HP and HPX
Filtri Cartucce con filtro
These cartridge filters have superior retention efficiency with rapid particle removal and microbubble control which facilitates faster bath startup and system qualification.
  • Ensures full compatibiliy and eliminates shedding that can occur after extended exposure
  • Patented hydrophilic membrane structure eliminates nucleation sites to reduce the potential for microbubble formation
Membrane: Patented UPE; Supports, core, sleeve and endcaps: HDPE
Maximum operating temperature: 60 °C
Maximum forward/reverse differential pressure: 3,4 bar at 23 °C