Stampa…

Sulfathiazole

Fornitore: EHRENSTORFER

C17000200
EHERC17000200EA 98.5 EUR
EHERC17000200
Sulfathiazole
Sulfathiazole
Formula: C₉H₉N₃O₂S₂
MDL Number: MFCD00005319
CAS Number: 72-14-0
EINECS: 200-771-5

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR