Stampa…

Propargite

Fornitore: EHRENSTORFER

C16430000
EHERC16430000EA 136 EUR
EHERC16430000
Propargite
Propargite
Formula: C₁₉H₂₆O₄S
MDL Number: MFCD00072448
CAS Number: 2312-35-8
EINECS: 219-006-1

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR