Stampa…

1-Methyl-3-nitroguanidine

Fornitore: EHRENSTORFER

C15106500
EHERC15106500EA 118 EUR
EHERC15106500
1-Methyl-3-nitroguanidine
1-Methyl-3-nitroguanidine
Formula: C₂H₅N₅O₃
Peso molecolare: 147.09 g/mol
MDL Number: MFCD00024167
CAS Number: 4245-76-5

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR