Stampa…
TIC instrument controller, 3 head
Fornitore: EDWARDS HIGH VACUUM METTLER
D39700000
EDWAD39700000EA 0 EUR
EDWAD39700000
TIC instrument controller, 3 head
Pompe Vacuometri
TIC Instrument controller is a compact control with a large, clear graphical display, an intuitive user interface and serial communications.
  • Automatically recognises and controls active gauges, including APG100 Pirani, convection, thermocouple, strain, inverted magnetron, wide range and active ion gauges
  • Enables complete integration into PC and PLC controlled processes, all TIC variants include RS232 and RS485 interface
Windows™-based PC program provides full remote setup, control and data logging functions via the RS232 interface.
Informazioni perMore Product Information
Visualizza i codici
Specifiche
Environmentally Preferable
Temperatura ambiente consentita per il funzionamento 0...40 °C (measured underneath TIC)
Alimentazione 90 - 264 V AC, 47 - 63 Hz
Peso 45108 kg