Stampa…
Rotary vane pumps, RV models
Fornitore: EDWARDS HIGH VACUUM METTLER
A65201903 A65301903 A65401903 A65501903 A65309903
181-0358EA 11010 EUR
181-0358 181-0360 531-0018 181-0361 EDWAA65309903
Rotary vane pumps, RV models
Pompe Pompe per vuoto a ventola rotante
These high performance rotary pumps are used in many industrial and laboratory applications. They offer the option of high throughput or high ultimate vacuum by the adjustment of a single control. RV pumps can be run continuosly with inlet pressures up to 50 mbar or can be set to achieve an ultimate partial pressure of 1×10⁻⁴ mbar. For drying applications the water handling capability is 290 g per hour at vapour pressures up to 80 mbar (RV 12).
  • Smooth running and quiet (48 dB A at 1 mbar)
  • High vacuum or high throughput switch
  • Inlet and outlet fittings NW