Stampa…
Experimentation kits, whose fingerprints were left behind?
Fornitore: EDVOTEK
S-91
EDVOS-91EA 94.8 EUR
EDVOS-91
Experimentation kits, whose fingerprints were left behind?
Materiale didattico Materiali didattici forensi Kit per scienza forense
Evidence left behind at a crime scene can identify a potential culprit. Even in this age of DNA, fingerprints and blood stains are still important in helping to identify a criminal. In this experiment, students solve a crime by dusting for fingerprints and use fluorescent dust to search for and identify trace amounts of blood.
  • For 10 lab groups
  • Complete in 50 minutes
Also required: Alcohol and longwave UV light.
Informazioni per l'ordine: Kit includes instructions, brushes, magnifying lens, fingerprint cards, black dusting powder, fluorescent greenMore Product Information