Stampa…
Floating racks
Fornitore: EDVOTEK
689
EDVO689EA 22.8 EUR
EDVO689
Floating racks
Rack Rastrelliere per tubi
Floating rack is ideal for floating tubes in water baths and tubs.
Length: 11,43 cm
Width: 7,62 cm