Stampa…
Experimentation kits, ion exchange chromatography
Fornitore: EDVOTEK
243
EDVO243EA 162 EUR
EDVO243
Experimentation kits, ion exchange chromatography
Materiale didattico Materiali didattici di chimica Attività di apprendimento - Cromatografia
Most molecules have a net charge within a pH range of 2 to 10. When the pH is altered, the net charge on molecules can change drastically. In this experiment, a mixture of 2 chemicals is absorbed onto a solid support ion-exchange column and separated during elution under conditions that influence their net charge.
  • For 6 lab groups
  • Requires 60-90 minutes
Also required: Spectrophotometer and cuvettes, ring stands and clamps, and 5 ml pipettes.
Informazioni per l'ordine: Kit includes instructions, ion exchanger, chemical mixture, potassium acetateMore Product Information