Stampa…

Erythrosin B disodium salt, Sigma-Aldrich®

Fornitore: SIGMA ALDRICH MICROSCOPY
Attenzione

Synonyms: Erythrosina 239, C.I. 45430

MQ200

E8886-25G
E8886-25GEA 86.5 EUR
E8886-25G
Erythrosin B disodium salt, Sigma-Aldrich®
Erythrosin B disodium salt
Formula: C₂₀H₆I₄Na₂O₅
Peso molecolare: 879.86 g/mol
MDL Number: MFCD00144257
CAS Number: 16423-68-0
EINECS: 240-474-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4a3e993c2552b7010c219308590ca3f81d3d2142&vid=dd3cfbaa1919e7721ce2f8fb646167b9d903e943&prid=web&clid=SAPDAM

Visualizza i codici

Specifiche

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR