Stampa…
Alkaline detergent, Divoflow 50
Fornitore: Diversey
7511265
DIVY7511265EA 189 EUR
DIVY7511265
Alkaline detergent, Divoflow 50
Soluzioni detergenti Detergenti e Detersivi per Superfici
Divoflow 50 is a low foaming alkaline liquid detergent suitable for CIP, bottle washer and other spray applications. It effectively removes organic soils and prevents the formation of tartar.
  • Clear pale yellow liquid
  • Versatile
  • Provides excellent performance with hard water
Divoflow 50 is highly effective at removing a wide range of organic soils and preventing scale build up and can be used in all food and beverage applications, where high causticity and scale inhibition is required. Highly effective detergent for cleaning smoke chambers with lowMore Product Information