Stampa…
Tubing, perbunan rubber
Fornitore: DEUTSCH NEUMANN
3241217 3240812 3240408 3240610 3240711
228-2353EA 44.4 EUR
228-2353 DENE3240812 DENE3240408 DENE3240610 DENE3240711
Tubing, perbunan rubber
Tubazioni Tubazione di trasferimento
Acrylonitrile-Butadiene (NBR), black.
  • Hardness: 70 Shore A
  • Temperatur resistance: −30 to +80 °C
  • No inlay
  • Resistant against oil.
  • Dimension tolerances according to DIN ISO 3302-1 E2