Stampa…
Dropping pipettes
Fornitore: DELTALAB
201110
DELT201110EA 133 EUR
DELT201110
Dropping pipettes
Pipette Pipette Pasteur/di trasferimento
Made of transparent, inert LDPE.
  • Can be frozen in liquid nitrogen
  • Volumetric tolerance: ±10%
Ideal for use as volume measurers, to incorporate in kits of reagents, pregnancy tests.
Press the upper bulb before use, filling all the stem, and the rest remains in the inferior bulb.