Stampa…
Heat resistant gloves, Terry Heavy
Fornitore: Honeywell Safety
RQE9607A
DALLRQE9607AEA 648 EUR
DALLRQE9607A
Heat resistant gloves, Terry Heavy
Guanti
Knitted cotton gloves with winged thumb and long cotton cuff. Suitable for mechanical and thermal hazards up to 250 °C.
  • Maximum comfort and added sturdiness are ensured
  • 5-finger gloves for a better grip of hot objects as well as added dexterity
  • Cuff helps to protect the forearm from mechanical and thermal hazards
Ideal for handling in glassworks, metallurgy and foundries.
Certificazioni: EN 388:2016 (2X31C), EN 407 (X1XXXX), Cat. II.