Stampa…
Perfect Fit™ Cut Resistant Gloves
Fornitore: Honeywell Safety
NPF21-1118W-11
DALLNPF21-1118W-11EA 37.1 EUR
DALLNPF21-1118W-11
Perfect Fit™ Cut Resistant Gloves
Guanti
The Honeywell Perfect Fit™ gloves were created with comfort in mind while also giving premium protection. The Perfect Fit™ Gloves are designed to protect workers from cuts, abrasions, lacerations, and punctures while working around sharp and rough materials.
  • Low content level of dmf (<100)
  • Colour coded markings
  • Silicon-free option
  • Coating breathability
  • Glass-fiber free for comfort
  • Light weight and Washable (up to 5 cycles of washing)
  • High grip performance
Perfect Fit™ has the unique Honeywell colour coding system for ease of identification, weMore Product Information