Stampa…
Supelco® Organic Reference Standard, F.A.M.E. Mix C8 - C24
Fornitore: Merck
CRM18918
CRM18918EA 111 EUR
CRM18918
Supelco® Organic Reference Standard, F.A.M.E. Mix C8 - C24
Standard Standard organici
This certified reference material is useful for determining relative retention times and approximating response factors.
  • GHS signal: GHS07
Attenzione: P273 - P391 - P501