Stampa…
Starter kit, Lambda™ Plus, Corning
Fornitore: Corning
4069
CORN4069EA 886.58 EUR
CORN4069
Starter kit, Lambda™ Plus, Corning
Pipette Pipette automatiche
The Lambda plus starter kit is designed for laboratories which are about to replace their old set of pipettors, and for customers who want to get a new set of pipettors which cover the most commonly used volumes.
The Lambda plus starter kit contains four Lambda Plus Single-Channel pipettors conveniently packaged with useful accessories (manual, calibration key, colour-coded rings, tube of piston grease and identification stickers).
Informazioni per l'ordine: This starter kit includes four Lambda Plus Single Channel pipettors in these volume sizes 0,5 to 10 µl, 2,0 to 20 µl, 20 to 200 µMore Product Information