Stampa…
Cleaning swabs, sealed, CONSTIX® SP-8
Fornitore: CONTEC
SP-8
CONTSP-8EA 68.5 EUR
CONTSP-8
Cleaning swabs, sealed, CONSTIX® SP-8
Tamponi Tamponi per la pulizia
The SP-8 sealed polyester swab consists of 100% knitted polyester fabric that is thermally bonded to a PP handle with a large paddle.
  • Durable and absorbent
  • Features extremely low particles and residue levels
  • Polyester head provides enhanced sorbency, particle entrapment and removal
  • Large paddle swab head provides broader surface coverage in critical applications
The swab head is made of cleanroom laundered; double-layer knitted microfibre polyester material. The polyester head is constructed without adhesives or binders, which makes the swabMore Product Information