Stampa…
Sealed foam swabs, SF-11 series, CONSTIX®
Fornitore: CONTEC
SF-11
CONTSF-11EA 238 EUR
CONTSF-11
Sealed foam swabs, SF-11 series, CONSTIX®
Tamponi Tamponi per la pulizia
CONSTIX® SF-11 swabs consist of a foam head formed over the end of a 152 mm cotton-tipped applicator with wood handle. The foam head is an 100ppi sealed closed-cell polyurethane foam.
  • Birch hardwood handle makes it stiff enough for vigorous swabbing
The soft, polyurethane foam head provides good particle entrapment and removal. The absorbent foam enables solvents, such as isopropanol and ethanol, to be readily absorbed for applications involving solvent delivery or removal. No adhesives are used, and the wood handle is solvent resistant. The swab isMore Product Information