Stampa…
Cleaning swabs, double-ended, CONSTIX® SC-2
Fornitore: CONTEC
SC-2
CONTSC-2EA 394 EUR
CONTSC-2
Cleaning swabs, double-ended, CONSTIX® SC-2
Tamponi Tamponi per la pulizia
SC-2 double-ended round cotton swabs consist of high-quality, long staple-length cotton, tightly wrapped and shaped on both ends of high strength, wound paper handles.
  • Medium, round cotton ends
  • High-quality cotton fibers provide excellent particle entrapment and retention
  • Tightly wrapped head help maintain low levels of extractable residue
  • Heads are held to the handles using a methyl-cellulose binder
The high-quality cotton fibers provide exceptional sorbency of most solvents and solutions. The heads are very tightly wrapped and shaped using a methyl-More Product Information