Stampa…

Anti-HA tag Rabbit Polyclonal Antibody (RPE (R-Phycoerythrin))

Fornitore: Columbia Biosciences
D5-1830
COBSD5-1830EA 629 EUR
COBSD5-1830
Anti-HA tag Rabbit Polyclonal Antibody (RPE (R-Phycoerythrin))
Anticorpi
Type: Tag
Antigen: HA tag (HA tag)
Clonality: polyclonal
Clone:
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR