Stampa…
Total Acid Number (TAN) Standards
Fornitore: CUSTOM MADE CHEMICALS LAB
TAN0001.075G TAN0020.075G TAN0100.075G TAN0500.075G
CMPLTAN0001.075GEA 0 EUR
CMPLTAN0001.075G CMPLTAN0020.075G CMPLTAN0100.075G CMPLTAN0500.075G
Total Acid Number (TAN) Standards
Standard Standard per l'industria peltrolchimica
Certificazioni: ISO 9001 certified, accredited according to ISO/IEC 17025 - Testing, accredited according to ISO 17034 - Reference Material Producer.
Informazioni per l'ordine: Shelf life on ship date: 24 Months - Store under normal laboratory conditions, at temperatures between 15 to 25 °C.